siRNA Details
| Gene Symbol: Mageb16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025013.1 | NP_001020184.1 | 3 | 781 | ||
| XM_006256976.1 | XP_006257038.1 | 4 | 841 | |||
| XM_006256977.2 | XP_006257039.1 | 5 | 904 | |||
| GenBank mRNA | BC079208.1 | - | 3 | 781 | 7131795 | |

Mageb16
Genomic Map