siRNA Details
| Gene Symbol: Mtmr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191725.2 | NP_001178654.1 | 11 | 1366 | ||
| XM_006254380.2 | XP_006254442.1 | 12 | 1458 | |||
| XM_006254381.2 | XP_006254443.1 | 12 | 1430 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr1| Gene Symbol: Mtmr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191725.2 | NP_001178654.1 | 11 | 1366 | ||
| XM_006254380.2 | XP_006254442.1 | 12 | 1458 | |||
| XM_006254381.2 | XP_006254443.1 | 12 | 1430 | |||