siRNA Details
| Gene Symbol: Ddx3x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108246.1 | NP_001101716.1 | 11 | 1217 | ||
| XM_006256652.2 | XP_006256714.1 | 13 | 1513 | |||
| XM_006256653.2 | XP_006256715.1 | 13 | 1510 | |||
| GenBank mRNA | BC085914.1 | - | 12 | 1558 | ||
| FQ211849.1 | - | 13 | 1492 | |||

Ddx3x
Genomic Map