siRNA Details
| Gene Symbol: RGD1565685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126286.2 | NP_001119758.2 | 3, 4 | 482 | ||
| XM_006256656.1 | XP_006256718.1 | 3, 4 | 2143 | |||
| XM_006256657.2 | XP_006256719.1 | 3, 4 | 2143 | |||
| XM_006256658.2 | XP_006256720.1 | 3, 4 | 2143 | |||
| GenBank mRNA | BC161948.1 | - | 3, 4 | 482 | 9025517 | |

RGD1565685
Genomic Map