siRNA Details
| Gene Symbol: Was | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108248.1 | NP_001101718.1 | 2 | 270 | ||
| GenBank mRNA | BC158677.1 | - | 2 | 286 | 8372629 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Was| Gene Symbol: Was | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108248.1 | NP_001101718.1 | 2 | 270 | ||
| GenBank mRNA | BC158677.1 | - | 2 | 286 | 8372629 | |