siRNA Details
| Gene Symbol: Tfe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271227.1 | NP_001258156.1 | 8 | 1358 | ||
| XM_006256721.2 | XP_006256783.1 | 8 | 2003 | |||
| XM_006256722.1 | XP_006256784.1 | 7 | 1248 | |||
| XM_006256723.2 | XP_006256785.1 | 8 | 1324 | |||
| XM_006256725.2 | XP_006256787.1 | 8 | 1017 | |||
| XM_006256726.2 | XP_006256788.1 | 8 | 967 | |||
| XM_006256727.2 | XP_006256789.1 | 8 | 1367 | |||
| XM_006256728.2 | XP_006256790.1 | 8 | 1036 | |||
| XM_006256729.2 | XP_006256791.1 | 8 | 987 | |||

Tfe3
Genomic Map