siRNA Details
| Gene Symbol: Tfe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271227.1 | NP_001258156.1 | 10 | 1736 | ||
| XM_006256721.2 | XP_006256783.1 | 10 | 2381 | |||
| XM_006256722.1 | XP_006256784.1 | 9 | 1626 | |||
| XM_006256723.2 | XP_006256785.1 | 10 | 1702 | |||
| XM_006256725.2 | XP_006256787.1 | 10 | 1395 | |||
| XM_006256726.2 | XP_006256788.1 | 10 | 1345 | |||
| XM_006256727.2 | XP_006256789.1 | 10 | 1745 | |||
| XM_006256728.2 | XP_006256790.1 | 10 | 1414 | |||
| XM_006256729.2 | XP_006256791.1 | 10 | 1365 | |||

Tfe3
Genomic Map