siRNA Details
| Gene Symbol: Foxp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108250.1 | NP_001101720.1 | 13 | 1471 | ||
| XM_006256731.2 | XP_006256793.1 | 12 | 1534 | |||
| GenBank mRNA | AB232988.1 | - | 13 | 1495 | ||
| AB232989.1 | - | 12 | 1413 | |||
| AB232990.1 | - | 12 | 1418 | |||

Foxp3
Genomic Map