siRNA Details
| Gene Symbol: Fus | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012137.1 | NP_001012137.1 | 3 | 229 | ||
| XM_006230290.2 | XP_006230352.1 | 3 | 256 | |||
| GenBank mRNA | BC087153.1 | - | 3 | 229 | 7132489 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fus| Gene Symbol: Fus | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012137.1 | NP_001012137.1 | 3 | 229 | ||
| XM_006230290.2 | XP_006230352.1 | 3 | 256 | |||
| GenBank mRNA | BC087153.1 | - | 3 | 229 | 7132489 | |