siRNA Details
| Gene Symbol: Ccnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773097.1 | XP_008771319.1 | 10 | 3951 | ||
| XM_008773098.1 | XP_008771320.1 | 10 | 3947 | |||
| XM_228779.6 | XP_228779.4 | 10 | 3948 | |||
| GenBank mRNA | HQ634292.1 | - | 9 | 3831 | ||

Ccnb3
Genomic Map