siRNA Details
| Gene Symbol: Shroom4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191730.1 | NP_001178659.1 | 6 | 3032 | ||
| XM_006256763.2 | XP_006256825.1 | 5 | 3109 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shroom4| Gene Symbol: Shroom4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191730.1 | NP_001178659.1 | 6 | 3032 | ||
| XM_006256763.2 | XP_006256825.1 | 5 | 3109 | |||