siRNA Details
| Gene Symbol: Ubqln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108251.1 | NP_001101721.1 | 1 | 1141 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_018798, BC053022, AK076159, AF177346, BC021824.

Ubqln2
Genomic Map