siRNA Details
| Gene Symbol: Gprasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257284.2 | XP_006257346.1 | 3150 | |||
| GenBank mRNA | FQ223397.1 | - | 3439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gprasp2| Gene Symbol: Gprasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257284.2 | XP_006257346.1 | 3150 | |||
| GenBank mRNA | FQ223397.1 | - | 3439 | |||