siRNA Details
| Gene Symbol: MGC109340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024267.2 | NP_001019438.1 | 1 | 457 | ||
| GenBank mRNA | BC092634.1 | - | 1 | 459 | 7315105 | |
| Gene Symbol: Arxes2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257283.2 | XP_006257345.1 | 1 | 751 | ||

MGC109340
Genomic Map