siRNA Details
| Gene Symbol: Phf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108253.1 | NP_001101723.1 | 7 | 964 | ||
| XM_006256792.2 | XP_006256854.1 | 7 | 1796 | |||
| XM_006256794.2 | XP_006256856.1 | 7 | 1204 | |||
| XM_006256795.2 | XP_006256857.1 | 7 | 1796 | |||
| XM_006256796.2 | XP_006256858.1 | 7 | 1795 | |||
| XM_008773119.1 | XP_008771341.1 | 7 | 934 | |||

Phf8
Genomic Map