siRNA Details
| Gene Symbol: Phf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108253.1 | NP_001101723.1 | 11 | 1420 | ||
| XM_006256792.2 | XP_006256854.1 | 11 | 2252 | |||
| XM_006256794.2 | XP_006256856.1 | 11 | 1660 | |||
| XM_006256795.2 | XP_006256857.1 | 11 | 2252 | |||
| XM_006256796.2 | XP_006256858.1 | 11 | 2251 | |||
| XM_008773119.1 | XP_008771341.1 | 11 | 1390 | |||

Phf8
Genomic Map