siRNA Details
| Gene Symbol: ART4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021071.2 | NP_066549.2 | 3 | 1247 | ||
| GenBank mRNA | AF290204.1 | - | 3 | 980 | ||
| AK291662.1 | - | 3 | 1263 | |||
| AY690666.1 | - | 2 | 100 | |||
| AY829431.1 | - | 3 | 193 | |||
| AY829432.1 | - | 2 | 121 | |||
| AY829434.1 | - | 3 | 1216 | |||
| AY829435.1 | - | 1 | 31 | |||
| BC074727.2 | - | 3 | 917 | 30915180 | ||
| BX096478.1 | - | 2 | 635 | |||

ART4
Genomic Map