siRNA Details
| Gene Symbol: Wwc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_228858.8 | XP_228858.8 | 4 | 905 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wwc3| Gene Symbol: Wwc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_228858.8 | XP_228858.8 | 4 | 905 | ||