siRNA Details
| Gene Symbol: Hccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191732.1 | NP_001178661.1 | 5 | 696 | ||
| XM_006256834.2 | XP_006256896.1 | 5 | 670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hccs| Gene Symbol: Hccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191732.1 | NP_001178661.1 | 5 | 696 | ||
| XM_006256834.2 | XP_006256896.1 | 5 | 670 | |||