siRNA Details
| Gene Symbol: LOC317456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047894.1 | NP_001041359.1 | 2 | 556 | ||
| GenBank mRNA | AY383668.1 | - | 2 | 556 | ||
| Gene Symbol: Lpgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109376.2 | NP_001102846.1 | 5, 6 | 955 | ||
| XM_008769875.1 | XP_008768097.1 | 6, 7 | 1013 | |||

LOC317456
Genomic Map