siRNA Details
| Gene Symbol: LOC317456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047894.1 | NP_001041359.1 | 1, 2 | 52 | ||
| GenBank mRNA | AY383668.1 | - | 1, 2 | 52 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC317456| Gene Symbol: LOC317456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047894.1 | NP_001041359.1 | 1, 2 | 52 | ||
| GenBank mRNA | AY383668.1 | - | 1, 2 | 52 | ||