siRNA Details
| Gene Symbol: Egfl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773171.1 | XP_008771393.1 | 10, 11 | 1529 | ||
| XM_008773172.1 | XP_008771394.1 | 10, 11 | 1526 | |||
| GenBank mRNA | BC081944.1 | - | 10, 11 | 1455 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Egfl6| Gene Symbol: Egfl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773171.1 | XP_008771393.1 | 10, 11 | 1529 | ||
| XM_008773172.1 | XP_008771394.1 | 10, 11 | 1526 | |||
| GenBank mRNA | BC081944.1 | - | 10, 11 | 1455 | ||