siRNA Details
| Gene Symbol: LOC317471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014112.2 | NP_001014134.2 | 1 | 1357 | ||
| GenBank mRNA | BC078965.1 | - | 1 | 1357 | 7111432 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC317471| Gene Symbol: LOC317471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014112.2 | NP_001014134.2 | 1 | 1357 | ||
| GenBank mRNA | BC078965.1 | - | 1 | 1357 | 7111432 | |