siRNA Details
| Gene Symbol: Nhs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191733.1 | NP_001178662.1 | 8 | 5174 | ||
| XM_008773173.1 | XP_008771395.1 | 9 | 5638 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nhs| Gene Symbol: Nhs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191733.1 | NP_001178662.1 | 8 | 5174 | ||
| XM_008773173.1 | XP_008771395.1 | 9 | 5638 | |||