siRNA Details
| Gene Symbol: Ppef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034935.1 | NP_001030107.1 | 3 | 293 | ||
| GenBank mRNA | BC104696.1 | - | 3 | 293 | 7112183 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppef1| Gene Symbol: Ppef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034935.1 | NP_001030107.1 | 3 | 293 | ||
| GenBank mRNA | BC104696.1 | - | 3 | 293 | 7112183 | |