siRNA Details
| Gene Symbol: Il1rapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177935.2 | NP_808796.1 | 9 | 1234 | ||
| XM_008773261.1 | XP_008771483.1 | 4 | 994 | |||
| GenBank mRNA | AY216593.1 | - | 9 | 1234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1rapl1| Gene Symbol: Il1rapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177935.2 | NP_808796.1 | 9 | 1234 | ||
| XM_008773261.1 | XP_008771483.1 | 4 | 994 | |||
| GenBank mRNA | AY216593.1 | - | 9 | 1234 | ||