siRNA Details
| Gene Symbol: Smarca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271190.1 | NP_001258119.1 | 5 | 733 | ||
| XM_006257525.2 | XP_006257587.2 | 6 | 735 | |||
| XM_006257526.2 | XP_006257588.2 | 6 | 750 | |||
| XM_006257527.2 | XP_006257589.2 | 6 | 750 | |||
| XM_008773524.1 | XP_008771746.1 | 6 | 750 | |||
| XM_008773526.1 | XP_008771748.1 | 6 | 703 | |||

Smarca1
Genomic Map