siRNA Details
| Gene Symbol: Sash3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134992.1 | NP_001128464.1 | 6 | 823 | ||
| XM_006257530.2 | XP_006257592.1 | 6 | 877 | |||
| GenBank mRNA | BC169091.1 | - | 6 | 823 | 7933144 | |
| FQ226243.1 | - | 2 | 125 | |||

Sash3
Genomic Map