siRNA Details
| Gene Symbol: Sash3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134992.1 | NP_001128464.1 | 2 | 286 | ||
| XM_006257530.2 | XP_006257592.1 | 2 | 340 | |||
| GenBank mRNA | BC169091.1 | - | 2 | 286 | 7933144 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sash3| Gene Symbol: Sash3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134992.1 | NP_001128464.1 | 2 | 286 | ||
| XM_006257530.2 | XP_006257592.1 | 2 | 340 | |||
| GenBank mRNA | BC169091.1 | - | 2 | 286 | 7933144 | |