siRNA Details
| Gene Symbol: Utp14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014113.2 | NP_001014135.2 | 14 | 2025 | ||
| GenBank mRNA | BC088326.1 | - | 4 | 1185 | 7365084 | |
| BC099799.1 | - | 14 | 2025 | 7936638 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Utp14a| Gene Symbol: Utp14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014113.2 | NP_001014135.2 | 14 | 2025 | ||
| GenBank mRNA | BC088326.1 | - | 4 | 1185 | 7365084 | |
| BC099799.1 | - | 14 | 2025 | 7936638 | ||