siRNA Details
| Gene Symbol: Stk26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191736.1 | NP_001178665.1 | 11 | 1419 | ||
| XM_006257573.2 | XP_006257635.1 | 13 | 1906 | |||
| XM_008773567.1 | XP_008771789.1 | 12 | 2618 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk26| Gene Symbol: Stk26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191736.1 | NP_001178665.1 | 11 | 1419 | ||
| XM_006257573.2 | XP_006257635.1 | 13 | 1906 | |||
| XM_008773567.1 | XP_008771789.1 | 12 | 2618 | |||