siRNA Details
| Gene Symbol: Magec2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025713.1 | NP_001020884.1 | 4 | 276 | ||
| XM_008773568.1 | XP_008771790.1 | 4 | 257 | |||
| GenBank mRNA | BC097464.1 | - | 4 | 276 | 7457620 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magec2| Gene Symbol: Magec2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025713.1 | NP_001020884.1 | 4 | 276 | ||
| XM_008773568.1 | XP_008771790.1 | 4 | 257 | |||
| GenBank mRNA | BC097464.1 | - | 4 | 276 | 7457620 | |