siRNA Details
| Gene Symbol: Map7d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271327.1 | NP_001258256.1 | 15 | 2333 | ||
| XM_008773649.1 | XP_008771871.1 | 15 | 2505 | |||
| XM_008773650.1 | XP_008771872.1 | 16 | 2471 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map7d3| Gene Symbol: Map7d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271327.1 | NP_001258256.1 | 15 | 2333 | ||
| XM_008773649.1 | XP_008771871.1 | 15 | 2505 | |||
| XM_008773650.1 | XP_008771872.1 | 16 | 2471 | |||