siRNA Details
| Gene Symbol: Nsa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134415.2 | NP_599242.2 | 6 | 825 | ||
| GenBank mRNA | BC098657.1 | - | 6 | 816 | 7378282 | |
| BP486932.1 | - | 3 | 320 | |||
| DQ354588.1 | - | 6 | 815 | |||
| FQ214845.1 | - | 6 | 843 | |||
| FQ225571.1 | - | 6 | 838 | |||
| FQ226635.1 | - | 6 | 852 | |||
| FQ229542.1 | - | 6 | 843 | |||
| Y17325.1 | - | 6 | 796 | |||

Nsa2
Genomic Map