siRNA Details
| Gene Symbol: Baz2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108260.1 | NP_001101730.1 | 11 | 2069 | ||
| XM_008761847.1 | XP_008760069.1 | 9 | 1621 | |||
| XM_008761848.1 | XP_008760070.1 | 8 | 1579 | |||
| XM_008761849.1 | XP_008760071.1 | 9 | 1615 | |||
| XM_008761850.1 | XP_008760072.1 | 9 | 1621 | |||
| XM_008761851.1 | XP_008760073.1 | 9 | 1621 | |||
| XM_008761852.1 | XP_008760074.1 | 9 | 1615 | |||
| XM_008761853.1 | XP_008760075.1 | 9 | 1621 | |||
| XM_008761854.1 | XP_008760076.1 | 9 | 1615 | |||
| XM_008761855.1 | XP_008760077.1 | 8 | 1432 | |||
| XM_008761856.1 | XP_008760078.1 | 7 | 1390 | |||
| XM_008761857.1 | XP_008760079.1 | 8 | 1426 | |||
| XM_008761858.1 | XP_008760080.1 | 9 | 1621 | |||

Baz2b
Genomic Map