siRNA Details
| Gene Symbol: Baz2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108260.1 | NP_001101730.1 | 35 | 6327 | ||
| XM_008761847.1 | XP_008760069.1 | 33 | 5885 | |||
| XM_008761848.1 | XP_008760070.1 | 32 | 5843 | |||
| XM_008761849.1 | XP_008760071.1 | 33 | 5879 | |||
| XM_008761850.1 | XP_008760072.1 | 32 | 5852 | |||
| XM_008761851.1 | XP_008760073.1 | 33 | 5780 | |||
| XM_008761852.1 | XP_008760074.1 | 33 | 5768 | |||
| XM_008761853.1 | XP_008760075.1 | 32 | 5747 | |||
| XM_008761854.1 | XP_008760076.1 | 32 | 5741 | |||
| XM_008761855.1 | XP_008760077.1 | 32 | 5696 | |||
| XM_008761856.1 | XP_008760078.1 | 31 | 5654 | |||
| XM_008761857.1 | XP_008760079.1 | 32 | 5690 | |||
| XM_008761858.1 | XP_008760080.1 | 32 | 5597 | |||

Baz2b
Genomic Map