siRNA Details
| Gene Symbol: Bbc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173837.2 | NP_776209.1 | 1744 | |||
| GenBank mRNA | AY157758.1 | - | 1744 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbc3| Gene Symbol: Bbc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173837.2 | NP_776209.1 | 1744 | |||
| GenBank mRNA | AY157758.1 | - | 1744 | |||