siRNA Details
| Gene Symbol: Lat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173840.1 | NP_776212.1 | 6, 7 | 305 | ||
| XM_006249160.2 | XP_006249222.1 | 7, 8 | 476 | |||
| XM_008769138.1 | XP_008767360.1 | 6, 7 | 758 | |||
| GenBank mRNA | AY170849.1 | - | 6, 7 | 305 | ||
| FQ231408.1 | - | 4, 5 | 165 | |||
| FQ235223.1 | - | 4, 5 | 165 | |||

Lat2
Genomic Map