siRNA Details
| Gene Symbol: Taar8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175601.1 | NP_783191.1 | 1 | 994 | ||
| XM_006227718.2 | XP_006227780.2 | 2 | 1060 | |||
| XM_006227719.2 | XP_006227781.1 | 2 | 1482 | |||
| XM_008758615.1 | XP_008756837.1 | 3 | 1640 | |||
More Information
Set Membership: |
|

Taar8b
Genomic Map