siRNA Details
| Gene Symbol: Crip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022501.1 | NP_071946.1 | 7 | 788 | ||
| XM_008764943.1 | XP_008763165.1 | 7 | 1052 | |||
| GenBank mRNA | BC061774.1 | - | 7 | 757 | 5621903 | |
| D17512.1 | - | 7 | 788 | |||
| FQ226314.1 | - | 7 | 796 | |||
| FQ227529.1 | - | 7 | 798 | |||
| FQ230331.1 | - | 7 | 798 | |||
| FQ233584.1 | - | 7 | 802 | |||

Crip2
Genomic Map