siRNA Details
| Gene Symbol: Gpbar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177936.1 | NP_808797.1 | 1 | 499 | ||
| XM_006245235.2 | XP_006245297.1 | 2 | 1511 | |||
| GenBank mRNA | AB086172.1 | - | 1 | 499 | ||
| AB089310.1 | - | 1 | 499 | |||
| JX534229.1 | - | 2 | 806 | |||
More Information
Set Membership: |
|

Gpbar1
Genomic Map