siRNA Details
Gene Symbol: Tlr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198131.1 | NP_937764.1 | 2 | 562 | ||
GenBank mRNA | AY191271.1 | - | 2 | 562 | ||
AY859725.1 | - | 1 | 466 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tlr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198131.1 | NP_937764.1 | 2 | 562 | ||
GenBank mRNA | AY191271.1 | - | 2 | 562 | ||
AY859725.1 | - | 1 | 466 |