siRNA Details
| Gene Symbol: MAST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181089.1 | NP_851603.1 | 2 | 95 | ||
| GenBank mRNA | AY227206.1 | - | 2 | 95 | ||
| AY227207.1 | - | 2 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAST1| Gene Symbol: MAST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181089.1 | NP_851603.1 | 2 | 95 | ||
| GenBank mRNA | AY227206.1 | - | 2 | 95 | ||
| AY227207.1 | - | 2 | 95 | |||