siRNA Details
| Gene Symbol: Zbtb16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013181.1 | NP_001013199.1 | 2 | 1155 | ||
| GenBank mRNA | AB568266.1 | - | 1 | 817 | ||
| AY781102.1 | - | 2 | 1155 | |||
| AY781103.1 | - | 2 | 1155 | |||
| AY781104.1 | - | 2 | 1146 | |||

Zbtb16
Genomic Map