siRNA Details
| Gene Symbol: Zar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181385.2 | NP_852050.1 | 1 | 182 | ||
| GenBank mRNA | AY283175.1 | - | 1 | 182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zar1| Gene Symbol: Zar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181385.2 | NP_852050.1 | 1 | 182 | ||
| GenBank mRNA | AY283175.1 | - | 1 | 182 | ||