siRNA Details
| Gene Symbol: Sgms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181386.2 | NP_852051.2 | 6 | 1108 | ||
| GenBank mRNA | AY280961.1 | - | 2 | 691 | ||
| BC078690.1 | - | 1 | 375 | 7113038 | ||
| BC081693.1 | - | 6 | 1108 | 7132545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgms1| Gene Symbol: Sgms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181386.2 | NP_852051.2 | 6 | 1108 | ||
| GenBank mRNA | AY280961.1 | - | 2 | 691 | ||
| BC078690.1 | - | 1 | 375 | 7113038 | ||
| BC081693.1 | - | 6 | 1108 | 7132545 | ||