siRNA Details
| Gene Symbol: Nadsyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181480.1 | NP_852145.1 | 10 | 855 | ||
| GenBank mRNA | AB090805.1 | - | 10 | 855 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nadsyn1| Gene Symbol: Nadsyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181480.1 | NP_852145.1 | 10 | 855 | ||
| GenBank mRNA | AB090805.1 | - | 10 | 855 | ||