siRNA Details
Gene Symbol: Gtf2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001512.2 | NP_001001512.2 | 22 | 2173 | ||
XM_006249162.2 | XP_006249224.1 | 26 | 2438 | |||
XM_006249163.2 | XP_006249225.1 | 26 | 2435 | |||
XM_006249164.2 | XP_006249226.1 | 25 | 2378 | |||
XM_006249165.2 | XP_006249227.1 | 25 | 2375 | |||
XM_006249166.2 | XP_006249228.1 | 25 | 2372 | |||
XM_006249167.2 | XP_006249229.1 | 24 | 2321 | |||
XM_006249168.2 | XP_006249230.1 | 24 | 2315 | |||
XM_006249169.2 | XP_006249231.1 | 24 | 2300 | |||
XM_006249170.2 | XP_006249232.1 | 26 | 2633 | |||
XM_006249171.2 | XP_006249233.1 | 23 | 2258 | |||
XM_006249172.2 | XP_006249234.1 | 23 | 2243 | |||
XM_006249173.2 | XP_006249235.1 | 23 | 2237 | |||
XM_006249174.2 | XP_006249236.1 | 22 | 2180 | |||
XM_008769141.1 | XP_008767363.1 | 26 | 2300 | |||
GenBank mRNA | AF547390.1 | - | 24 | 2210 | ||
AF547391.1 | - | 24 | 2210 | |||
BC085815.1 | - | 22 | 2311 | 7123295 |
