siRNA Details
| Gene Symbol: Col23a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181636.2 | NP_853667.1 | 1029 | |||
| GenBank mRNA | AY158896.1 | - | 1029 | |||
| Gene Symbol: LOC103693323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767739.1 | XP_008765961.1 | 335 | |||

Col23a1
Genomic Map