siRNA Details
| Gene Symbol: Pigu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181637.1 | NP_853668.1 | 3 | 250 | ||
| XM_006235325.2 | XP_006235387.1 | 4 | 501 | |||
| XM_008762347.1 | XP_008760569.1 | 1 | 598 | |||
| GenBank mRNA | AB086841.1 | - | 3 | 285 | ||
| BC097269.1 | - | 4 | 416 | |||

Pigu
Genomic Map